Merck
14C-labeling of bromobutide, 2-bromo-3,3-dimethyl-N-(alpha, alpha-dimethylbenzyl)butyramide.
Radioisotopes (1988-01-01)
H Kanamaru, K Nishioka, H Komori, K Kawahara
ABSTRAKT
Bromobutide, a novel herbicide, was labeled with carbon-14 independently at the carbonyl group and the phenyl ring for use in metabolic studies. 14C-Carbonation of neopentylmagnesium chloride (3) gave 3,3-dimethyl[1-14C]butyric acid (4a) quantitatively. Chlorination of 4a with thionyl chloride followed by alpha-bromination with bromine yielded 2-bromo-3,3-dimethyl[1-14C]-butyryl halide (5a), which was subsequently condensed with alpha, alpha-dimethylbenzylamine (6a) to afford [carbonyl-14C]bromobutide (1a). The overall yield of 1a was 76% from barium [14C]-carbonate (2). Similarly, condensation of alpha, alpha-dimethyl[phenyl-14C]benzylamine (6b), which was prepared from alpha-methyl[phenyl-U-14C]styrene (7) in three steps, with 2-bromo-3,3-dimethylbutyryl halide (5b) gave [phenyl-14C]bromobutide (1b) in 67% yield after purification. The specific activities of 1a and 1b were 1.38 and 0.781 GBq/mmol (37.2 and 21.1 mCi/mmol), respectively.