- [Microdetermination of pyrazinamide and its metabolites (pyrazinoic acid, 5-hydroxypyrazinoic acid, 5-hydroxypyrazinamide and pyrazinuric acid) in plasma and urine with liquid chromatography].
[Microdetermination of pyrazinamide and its metabolites (pyrazinoic acid, 5-hydroxypyrazinoic acid, 5-hydroxypyrazinamide and pyrazinuric acid) in plasma and urine with liquid chromatography].
Journal of chromatography (1987-11-27)
C Lacroix, P Poncet, G Laine, C Guyonnaud, M Ray, S Menager, O Lafont
PMID3437007
ABSTRAKT
The method reported here for determining pyrazinamide and its metabolites (2-pyrazinoic acid, 5-hydroxypyrazinamide, 5-hydroxypyrazinoic acid and pyrazinuric acid) consists of diluting urine or acid deproteinisation of serum followed by chromatography on a cation-exchange column. The column length and the detection system (ultraviolet or fluorimetry) allow for a very good separation of the different compounds; the sensitivity of the method makes it suitable for pharmacokinetic studies.