Merck
Metabolism of monofluoro- and monochlorobenzoates by a dentrifying bacterium.
Archives of microbiology (1979-09-01)
B F Taylor, W L Hearn, S Pincus
PMID543775
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3-Fluorobenzoic acid, 97%
Sigma-Aldrich
2-Fluorobenzoic acid, 97%