Merck
[Cresol burns in the eye (author's transl)].
Klinische Monatsblatter fur Augenheilkunde (1979-10-01)
E Schütte, B Hörster, R Schäfer
PMID548623
ABSTRAKT
A brief introduction to the pathomechanism of erosions in the eye is followed by a case report of a young soldier injured by a "peptizer" (2-methoxy-4-methylphenol). After early application of cortisone there was no cicatrization impairing function in the central corneal stroma. A row of small vesicles at the corneal limbus and in the conjunctiva were reminiscent of Labrador corneal dystrophy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Methoxy-4-methylphenol, ≥98%
Sigma-Aldrich
2-Methoxy-4-methylphenol, ≥98%, FG
Sigma-Aldrich
2-Methoxy-4-methylphenol, natural, 97%, FG