Merck
[Antiviral activity of chelidamic acid derivatives].
Voprosy virusologii (1984-05-01)
V E Iavorovskaia, V I Il'enko, G N Tiurenkova, N V Serebriakova, A N Evstropov
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chelidamic acid hydrate, ≥95%, powder
Sigma-Aldrich
Chelidamic acid hydrate, ≥97.0% (dried material, T), ~1 mol/mol water