Merck
Effect of pyridines on phenotypic properties of Bordetella pertussis.
Infection and immunity (1982-11-01)
D R Schneider, C D Parker
ABSTRAKT
Several conditions of growth of Bordetella pertussis cause a reversible phenotypic alteration in properties termed modulation. Growth in medium containing nicotinic acid induces normal (X-mode) cells to change to modulated (C-mode) cells. We examined several pyridines and compounds resembling pyridines for their ability to affect modulation, using envelope protein patterns and serological reactivity as indicators of modulation. We found that 6-chloronicotinic acid and quinaldic acid were more effective modulating stimuli than was nicotinic acid on a molar basis. Both 2-chloronicotinamide and isoniazid interfered with nicotinic acid-induced modulation, and can be called antimodulators. Picolinic acid inhibited growth.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Quinaldic acid, 98%
Sigma-Aldrich
6-Chloropyridine-3-carboxylic acid, 99%