Merck
Interaction between newer cephalosporins and Clinitest, Diastix, and Tes-Tape.
American journal of hospital pharmacy (1983-09-01)
M C McManus, S L Barriere
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Benedict′s reagent, for the qualitative determination of sugars