Merck
[Health standardization of Cyanox in water].
Gigiena i sanitariia (1983-09-01)
I I Datsenko, E I Tolmacheva
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Cyanophos, PESTANAL®, analytical standard