- [Histochemical study of peroxidase and tyrosinase activities in the cuticle of Protophormia terraenovae].
[Histochemical study of peroxidase and tyrosinase activities in the cuticle of Protophormia terraenovae].
Angewandte Parasitologie (1980-02-01)
J Brunner, U Groth
PMID6769371
ABSTRAKT
Quantitative histochemical measurements of activities of enzymes were performed by temporal defined stages in development of Protophormia terraenovae. The effect of the specific insecticide of moult Duphar pH 60-38 activity of enzyme was evaluated. Activities of peroxidase and tyrosinase showed parallel oscillations, at which proof of both enzymes was restricted to the cuticle. The insects, treated with insecticide, showed a moulting, dependent on concentration. A transfer of maxima of activities is described.
MATERIAŁY