Merck
  • Synthesis of Bz-Phe(p-No2)-Val-Arg-pNA-and its specificity to thrombin-like enzymes: similarity of a commercial product S-2160 to the nitrated compound.
Synthesis of Bz-Phe(p-No2)-Val-Arg-pNA-and its specificity to thrombin-like enzymes: similarity of a commercial product S-2160 to the nitrated compound.
Thrombosis research (1980-12-01)
K Takada, S Sakakibara, H Kato, T Goto, S Iwanaga
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N-Benzoyl-Phe-Val-Arg-p-nitroanilide hydrochloride, protease substrate