Merck
  • Amino acids and peptides. XXXI. Phosphorus in organic synthesis. XVIII. Synthesis of porcine motilin by the solid-phase method using diphenyl phosphorazidate(DPPA) and diethyl phosphorocyanidate(DEPC).
Amino acids and peptides. XXXI. Phosphorus in organic synthesis. XVIII. Synthesis of porcine motilin by the solid-phase method using diphenyl phosphorazidate(DPPA) and diethyl phosphorocyanidate(DEPC).
Chemical & pharmaceutical bulletin (1980-11-01)
N Ikota, T Shioiri, S Yamada, S Tachibana
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Diphenyl phosphoryl azide, 97%