Merck
An improved gas chromatographic method for the detection and quantitation of mercaptans in blood.
Clinica chimica acta; international journal of clinical chemistry (1981-06-02)
H A Mardini, K Bartlett, C O Record
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Propyl mercaptan, ≥97%
Sigma-Aldrich
1-Propanethiol, 99%