Merck
Benzophenone metabolism. I. Isolation of p-hydroxybenzophenone from rat urine.
Life sciences (1980-02-04)
A W Stocklinski, O B Ware, T J Oberst
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Hydroxybenzophenone, 98%