Merck
Hydroxylations of biphenyl by fungi.
The Journal of applied bacteriology (1980-08-01)
R V Smith, P J Davis, A M Clark, S Glover-Milton
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4,4′-Dihydroxybiphenyl, 97%