Merck
Janus Green B as a rapid, vital stain for peripheral nerves and chordotonal organs in insects.
Journal of neuroscience methods (1993-08-01)
J E Yack
ABSTRAKT
Effective staining of peripheral nerves in live insects is achieved with the vital stain Janus Green B. A working solution of 0.02% Janus Green B in saline is briefly applied to the exposed peripheral nervous system. The stain is then decanted and the dissection flooded with fresh saline, resulting in whole nerves being stained dark blue in contrast to surrounding tissues. This simple and reliable technique is useful in describing the distribution of nerves to their peripheral innervation sites, and in locating small nerve branches for extracellular physiological recordings. The stain is also shown to be useful as a means of enhancing the contrast between scolopale caps and surrounding tissues in chordotonal organs, staining chordotonal organ attachment strands, and the crista acustica (tympanal organ) of crickets and katydids. The advantages of Janus Green B over traditional peripheral nerve strains, in addition to its shortcomings, are discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Janus Green B, certified by the BSC, Dye content 65 %