Merck
Pressure effects on enzyme reactions in mainly organic media: alpha-chymotrypsin in reversed micelles of Aerosol OT in octane.
Biochemistry and molecular biology international (1994-08-01)
V V Mozhaev, N Bec, C Balny
ABSTRAKT
Biocatalytic transformations in reversed micelles formed by anionic surfactant Aerosol OT in octane have been studied at high pressures by an example of alpha-chymotrypsin-catalyzed hydrolysis of N-carbobenzoxy-L-tyrosine p-nitrophenyl ester and N-succinyl-L-phenylalanine p-nitroanilide. For the first time it has been found that the enzyme retains high activity in these water-in-oil microemulsions up to a pressure of 2 kbar. The value of the activation volume (delta V*) for the enzyme reactions shows a dependence on the water content in the system. When the size of the micellar aqueous inner cavity (as evaluated at 1 atm) approaches the molecular size of alpha-chymotrypsin, delta V* becomes significantly different from the value in aqueous solution and in the micelles with a larger size. Possibilities of regulating the enzyme activity by pressure in systems with a low content of water are discussed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N-Succinyl-L-phenylalanine-p-nitroanilide, protease substrate
Informacje o cenach i dostępności nie są obecnie dostępne.