Merck
[A multichannel microelectrode system for long-term use].
Fiziologicheskii zhurnal imeni I.M. Sechenova (1994-08-01)
I F Kalmykov, A A Orlov
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Silanization solution I, ~5% (dimethyldichlorosilane in heptane), Selectophore
Sigma-Aldrich
Dichlorodimethylsilane, ≥99.5%
Sigma-Aldrich
Dichlorodimethylsilane, ≥98.5% (GC)
Sigma-Aldrich
Dichlorodimethylsilane, Wacker Chemie AG, ≥99.0% (GC)