Merck
  • Analysis of the neurotoxic plasticizer n-butylbenzenesulfonamide by gas chromatography combined with accurate mass selected ion monitoring.
Analysis of the neurotoxic plasticizer n-butylbenzenesulfonamide by gas chromatography combined with accurate mass selected ion monitoring.
Journal of analytical toxicology (1994-11-01)
P Duffield, D Bourne, K Tan, R M Garruto, M W Duncan
ABSTRAKT
The plasticizer, n-butylbenzenesulfonamide (NBBS), is reported to be neurotoxic when inoculated intracisternally or intraperitoneally into rabbits. Because NBBS is commonly used in the production of polyamide (nylon) plastics and is soluble in water, the disposal of NBBS-containing plastics in landfill sites could result in NBBS appearing in the leachate. Further, NBBS could also be leached from packaging into their contents. To allow us to examine the risks posed by NBBS in the environment, we have developed a quantitative assay for this compound. The assay employs a one-step extraction into dichloromethane followed by gas chromatography with accurate mass selected ion recording. The assay incorporates [13C6]NBBS as an internal standard to allow precise quantitation, and four separate ion chromatograms are recorded. NBBS was found in some Australian domestic solidwaste landfill leachate (from less than 0.3 to 94.6 ng/mL), but ground water in the vicinity of a landfill had only trace quantities of NBBS. NBBS was also quantitated in some bottled and cask wines, and levels varied from not detected to 2.17 ng/mL (n = 14). Additional studies are required to assess the public health risks associated with the use of NBBS as a plasticizer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N-Butylbenzenesulfonamide, 99%