Merck
  • [Analogs of 3'-dephospho-CoASH with a phosphodiester bond as substrates of the S-acetylation reaction, catalyzed by acetyl-CoA-synthetase fom rabbit myocardium].
[Analogs of 3'-dephospho-CoASH with a phosphodiester bond as substrates of the S-acetylation reaction, catalyzed by acetyl-CoA-synthetase fom rabbit myocardium].
Bioorganicheskaia khimiia (1993-09-01)
A I Biriukov, Iu N Zhukov, V M Kopelevich, L N Bulanova, V I Gunar
ABSTRAKT
A number of earlier unknown 3'-dephospho-CoASH analogues with the pyrophosphate fragment replaced by an ester or phosphodiester bond were synthesized and tested in S-acetylation reaction, catalyzed by acetyl-CoA synthetase (EC 6.2.1.1) from rabbit myocardium. 3'-Dephospho-CoASH analogues with a phosphodiester bond, e.g. (Ia), had a lower affinity and diminished kinetic parameters than 3'-dephospho-CoASH (Km = 1 and 0.2 mM, respectively). The adenine substitution in (Ia) by guanine or hypoxanthine (but not cytosine) residue resulted in a loss of substrate properties. 3'-Dephospho-CoASH with an ester bond were not capable of accepting acetate under conditions used and only slightly inhibited the enzymic activity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3′-Dephosphocoenzyme A, ≥90% (HPLC)