Merck
[The use of an impulse magnetic field in the combined therapy of patients with stone fragments in the upper urinary tract].
Voprosy kurortologii, fizioterapii, i lechebnoi fizicheskoi kultury (1994-05-01)
A A Li, N I Nesterov, S N Malikova, V A Kiiatkin
ABSTRAKT
The paper covers the experiment's details and the results obtained at stimulation of the kidney projection region and upper ureter with remote-controlled impulse magnetic field (IMF). It was found that IMF activates impulse activity of ureteral smooth muscles in 100% of cases. The technique is detailed of managing ureterolith fragments using IMF combined with iodobromine baths. The results of the proposed treatment are given. A single therapeutic course produced effect in 62.9% of the patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Iodine monobromide, 98%