Merck
The reaction sites of rotenone and ubiquinone with mitochondrial NADH dehydrogenase.
Biochimica et biophysica acta (1994-08-30)
T P Singer, R R Ramsay
ABSTRAKT
This article summarizes recent studies in the authors' and other laboratories of selective inhibitors acting at the 'rotenone' site and at the Q binding site in the NADH-Q oxidoreductase segment of the respiratory chain. A wide array of inhibitors act at the rotenone site to block electron flux from the enzyme to the Q pool. Using evidence from studies with rotenone, piericidin A, and analogs of the neurotoxic N-methyl-4-phenylpyridinium, we have proposed two binding sites for these inhibitors, both of which must be occupied for complete inhibition of NADH oxidation.