- [A case of allergic reaction to the sting by the bee].
[A case of allergic reaction to the sting by the bee].
Otolaryngologia polska = The Polish otolaryngology (1993-01-01)
J Zawadowski, E Zawisza
PMID8164967
ABSTRAKT
The authors described a case of an insect sting allergy, of a 19 year old patient. They discuss the treatment and prevention in such cases and situations.
MATERIAŁY