Merck
Reduction of disulfide bonds in proteins by 2-aminothiophenol under weakly acidic conditions.
Journal of biochemistry (1994-01-01)
Y Abe, T Ueda, T Imoto
ABSTRAKT
We developed a method for reducing disulfide bonds in proteins under weakly acidic conditions by use of 2-aminothiophenol. The disulfide bonds in hen egg-white lysozyme, ribonuclease A, and soybean trypsin inhibitor were quantitatively reduced by 2-aminothiophenol in phosphate buffer, pH6, containing 8 M Gdn HCl, 1 mM EDTA, and 20% ethanol, for 60 min at 40 degrees C. On analysis of the RP-HPLC patterns of tryptic peptides, which were derived from reduced and S-alkylated lysozyme and ribonuclease A at pH 6, it was confirmed that no side reaction occurred. Moreover, the reduction under weakly acidic conditions was demonstrated to be applicable for the location of such a labile residue as O-acetylated tyrosine.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Aminothiophenol, technical grade, 90%
Sigma-Aldrich
2-Aminothiophenol, 99%