Merck
Allergic contact dermatitis from the fungicide bupirimate.
Contact dermatitis (1993-01-01)
J P McFadden, M Kinoulty, R J Rycroft
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Bupirimate, PESTANAL®, analytical standard