Merck
Introduction of amine groups on poly(ethylene) by plasma immobilization of a preadsorbed layer of decylamine hydrochloride.
Journal of biomaterials science. Polymer edition (1993-01-01)
J G Terlingen, L M Brenneisen, H T Super, A P Pijpers, A S Hoffman, J Feijen
ABSTRAKT
In order to introduce amine groups on poly(ethylene) (PE) surface, PE surfaces were preadsorbed with decylamine hydrochloride (DA.HCl) and subsequently treated with an argon plasma. It was shown by XPS (X-ray Photoelectron Spectroscopy), that approximately half of the preadsorbed (mono)layer was immobilized and that a substantial part (60-70%) of the incorporated nitrogen containing groups were amine groups. The availability of the surface amine groups for reactions was investigated by applying a gas phase reaction with 4-trifluoromethylbenzaldehyde and by a reductive methylation reaction in aqueous solution with 14C formaldehyde. A maximal number of reactive amine groups was found after a plasma treatment time of 2 s. The reductive methylation reaction was used to estimate the surface concentration of amine groups resulting in a typical surface concentration of 1 x 10(-6) mol/m2 after a plasma treatment time of 2 s.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Decylamine, 95%
Sigma-Aldrich
Decylamine, ≥99.0% (GC)