Merck
Direct analysis of Kryptofix 2.2.2 in 18FDG by gas chromatography using a nitrogen-selective detector.
Nuclear medicine and biology (1993-04-01)
R A Ferrieri, D J Schlyer, D L Alexoff, J S Fowler, A P Wolf
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane, 98%