Merck
Paint as another possible source of lead exposure in Saudi Arabia.
Bulletin of environmental contamination and toxicology (1995-09-01)
I al-Saleh, L Coate
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Petroleum ether, anhydrous