Merck
An essential tyrosine residue of Aspergillus polygalacturonase.
FEBS letters (1996-03-11)
E Stratilová, M Dzúrová, O Markovic, H Jörnvall
ABSTRAKT
Based on strict conservation of a tyrosine residue in 24 polygalacturonases, tyrosine modification was assessed in two different forms of the Aspergillus enzyme. The second subform was unknown in structure but submitted to sequence analysis and was found also to have the conserved tyrosine residue. Results of chemical modifications are consistent in showing inactivation of the proteins with all tyrosine-reactive agents tested, acetic anhydride, N-acetyl imidazole, and tetranitromethane. Furthermore, after acetylation, regeneration of enzyme activity was possible with hydroxylamine. Spectrophotometric pH titration showed that one accessible tyrosine residue is ionized at pH 9.3-9.5, whereas the remaining, masked residues are all ionized at pH 10.5. It is concluded that one tyrosine residue is catalytically important, in agreement with the inactivation and reactivation data, that this residue is accessible, and that it is likely to correspond to the strictly conserved residue observed in all forms.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Acetylimidazole, 98%