Merck
[Preparation and antiviral properties of Bacillus intermedius ribonuclease modified by chloranhydride of adamantanecarboxylic acid].
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic] (1995-09-01)
B M Kurinenko, N V Kalacheva, P Iu Muratov
ABSTRAKT
Bacillus intermedius ribonuclease modified by the residue of adamantane carboxylic acid was prepared. When the cells of chick embryo fibroblasts infected by the fowl plague virus were exposed to the modified ribonuclease, the antiviral activity proved to be higher by comparison to that of the native enzyme. The chemotherapeutic index of the RNAse after the modification increased 4 times.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Adamantanecarboxylic acid, 99%