Merck
Rapid repigmentation after depigmentation therapy: vitiligo treated with monobenzyl ether of hydroquinone.
The Australasian journal of dermatology (1996-05-01)
A M Oakley
ABSTRAKT
Repigmentation occurred within a few weeks of discontinuing successful depigmentation therapy with monobenzyl ether of hydroquinone, in a patient with extensive vitiligo. Patients undertaking depigmentation therapy should be warned that this may occur. The mechanism is unknown.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-(Benzyloxy)phenol, 98%