Merck
The binding of cholesterol and bile salts to recombinant rat liver fatty acid-binding protein.
The Biochemical journal (1996-12-15)
A E Thumser, D C Wilton
ABSTRAKT
The physiological role of liver fatty acid-binding protein (L-FABP) has yet to be clarified. An important feature of this member of the family of intracellular lipid-binding proteins is the wide range of compounds that have been identified as potential physiological ligands. By using recombinant L-FABP, the binding of cholesterol, bile salts and their derivatives has been investigated under conditions that allow a direct comparison of the binding affinities of these ligands for fatty acids. The results demonstrate an inability of L-FABP to bind cholesterol, although the anionic derivative, cholesteryl sulphate, will bind under similar assay conditions. Of the bile salts examined, lithocholate and taurolithocholate sulphate showed the greatest binding to L-FABP. It is proposed that an important function of L-FABP is to bind certain physiological amphipathic anions, thus preventing the "free' concentrations of these compounds from exceeding their critical micelle concentration, which could result in cell damage.