- L-histidine methyl ester dihydrochloride.
L-histidine methyl ester dihydrochloride.
Acta crystallographica. Section C, Crystal structure communications (1996-03-15)
V H Vilchiz, R E Norman, S C Chang
PMID9004733
ABSTRAKT
The title compound, C7H13N3O2(2+).2Cl-, has distances and angles quite similar to those of histidine hydrochloride monohydrate [Donohue & Caron (1964). Acta Cryst. 17, 1178-1180], except for the distances within the ester functionality.
MATERIAŁY