Merck
  • The effects of 4-chloro-ortho-toluidine (fast red TR) subacute intoxication on rats. Part I: The kidney and the urinary bladder epithelium.
The effects of 4-chloro-ortho-toluidine (fast red TR) subacute intoxication on rats. Part I: The kidney and the urinary bladder epithelium.
Folia morphologica (1996-01-01)
A Godlewski, W Fortak, P M Brzeziński, J Dobek-Smiech, M J Stasik, J Konietzko
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fast Red TR Salt, Dye content 15 %