Merck
Spectrophotometric determination of the positional specificity of nonspecific and 1,3-specific lipases.
Analytical biochemistry (1997-11-05)
R N Farias, M Torres, R Canela
ABSTRAKT
Using commercially available thiosubstrates, such as 2,3-dimercapto-1-propanol tributyrate, the regio-specificities of 1,3-specific and nonspecific lipases was confirmed. The spectrophotometric test is a simple, rapid, and convenient alternative method to those previously reported for the characterization of the positional specificities of new lipases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2,3-Dimercapto-1-propanol tributyrate, 97%