Merck
Flow extraction spectrophotometric method for the determination of diclofenac sodium in pharmaceutical preparations.
Journal of pharmaceutical and biomedical analysis (1997-12-31)
T Pérez-Ruiz, C Martínez-Lozano, A Sanz, M T San Miguel
ABSTRAKT
The spectrophotometric determination of trace amounts of diclofenac was carried out by liquid-liquid extraction using acridine yellow with a flow system. The determination of diclofenac sodium in the range of 3-80 micrograms ml-1 was possible with a sampling frequency of 40 samples h-1. The method was satisfactorily applied to the determination of diclofenac in pharmaceutical preparations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acridine Yellow G, Dye content 90 %