Merck
Analysis of potentially toxic phthalate plasticizers used in toy manufacturing.
Bulletin of environmental contamination and toxicology (1998-03-04)
M L Marín, J López, A Sánchez, J Vilaplana, A Jiménez
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Diisodecyl phthalate, Selectophore, ≥99.0%