Merck
Cytochrome P-450 cholesterol side-chain cleavage expression in human spermatozoa.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme (1998-03-21)
R Gunasegaram, K L Peh, A Loganath, P C Chew, S S Ratnam, S Arulkumaran
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Methylpentanoic acid, ≥98%, FCC, FG
Sigma-Aldrich
4-Methylvaleric acid, 99%