Merck
Comparison of anion-exchange and ion-modified reversed-phase liquid chromatography for the determination of S-sulfocysteine.
Journal of chromatography. B, Biomedical sciences and applications (1998-04-01)
W Xia, M Sandberg, S G Weber
ABSTRAKT
A dual Hg-Au amalgam electrode is used to detect S-sulfocysteine (SSC) in this study. There exist two main components in the acetonitrile (ACN) rat brain extracts, namely, Cl- and GSSG (oxidized glutathione), that are active in our detection system (GSH is not extracted in ACN). Two strong anion-exchange columns from different companies were used to separate the samples under different conditions, but SSC and Cl- were not separated at the optimum detection pH of 5.2. The signal from Cl- was greatly decreased by lowering the potential at the downstream electrode, though it cannot be completely eliminated. While a silver cartridge removed Cl- from micromoles to several millimoles without any negative effect on the SSC signal in aqueous standards, a large negative peak which interferes with SSC detection was unfortunately introduced when a silver cartridge was applied to brain tissue samples. However, SSC and Cl- in the samples are successfully separated by ion-modified reversed-phase LC in acetate buffer at the optimum detection pH (5.2). The separation conditions are 20 mM acetic acid, 2% methanol, 0.5 mM cetyltrimethylammonium p-toluene sulfonate (CTMA) (pH 5.2). Most importantly, the sensitivity of SSC under the optimum separation conditions is not sacrificed. The detection limit is 8 nM (20 microl injected).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-Cysteine S-sulfate, ≥98% (TLC)