Merck
Contact hypersensitivity to dicyclohexylcarbodiimide and diisopropylcarbodiimide in female B6C3F1 mice.
Drug and chemical toxicology (1998-05-23)
B B Hayes, P C Gerber, S S Griffey, B J Meade
ABSTRAKT
Dicyclohexylcarbodiimide (DCC) and diisopropylcarbodiimide (DIC) are two commonly used coupling reagents in protein synthesis resulting in exposure of individuals in chemical and pharmaceutical industries as well as research laboratories involved in protein synthesis and recombinant DNA techniques. The objectives of these studies were to determine the irritation and sensitizing potential of these two compounds when applied topically to B6C3F1 mice. Sensitization potential was assessed by the Mouse Ear Swelling Test (MEST) and the murine Local Lymph Node Assay (LLNA). Concentrations used in the contact hypersensitivity assays were determined by primary irritancy studies. DCC and DIC were identified as both irritants and contact sensitizers with the MEST being a more sensitive indicator of sensitization potential. The MEST identified DCC as a sensitizer at concentrations as low as 0.006% (w/v) 24 hr and 48 hr post challenge and DIC at 0.3% (w/v) and 1.5% (w/v) 24 and 48 hr post challenge, respectively. In the LLNA, the lowest concentrations yielding a significant response were 0.06% (w/v) for DCC and 10% (w/v) for DIC.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DIC, 99%
Sigma-Aldrich
DIC, purum, ≥98.0% (GC)
Sigma-Aldrich
Diisopropylcarbodiimide solution, 1 M in THF