Merck
Isobutyl nitrite liberates nitric oxide which is not responsible for the immunotoxicity of the inhalant.
Advances in experimental medicine and biology (1998-07-17)
L S Soderberg, A Roy, J B Barnett
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Isobutyl nitrite, 95%