Merck
[Gas chromatographic determination of dicyclopentadiene in the work area air].
Gigiena i sanitariia (1998-11-17)
N V Shumilov, N M Ustinova
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dicyclopentadiene, contains BHT as stabilizer