Merck
Influence of reaction conditions on syntheses of sweetener precursors catalyzed by thermolysin in tert-amyl alcohol.
The journal of peptide research : official journal of the American Peptide Society (1998-12-01)
G W Xing, G L Tian, Y H Ye
ABSTRAKT
The activity of enzymes to form a peptide bond in organic solvent was greatly influenced by observed pH and water content. The precursors of two sweeteners, P-Asp-Xaa-OR (P=Z or For, Xaa-OR=Phe-OMe or Ala-OcHex), were synthesized by enzyme, and the reaction conditions were studied systematically. Z-Asp-OH was coupled with H-Phe-OMe or H-Ala-OcHex by thermolysin in tert-amyl alcohol. The best coupling results were obtained when the optimized observed pH was 8 or 9, and the water content was about 6% (V/V). The protecting group Z is better than For under the reaction conditions and H-Phe-OMe is a better nucleophile than H-Ala-OcHex. The expected optically pure peptides were obtained when the racemic amino acids were used as amino components in the starting materials. The physical constants of P-Asp-Xaa-OR synthesized by thermolysin are identical with those of peptides synthesized by chemical method.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Methyl-2-butanol, anhydrous, ≥99%
Sigma-Aldrich
2-Methyl-2-butanol, ReagentPlus®, ≥99%
Supelco
2-Methyl-2-butanol, analytical standard
Sigma-Aldrich
2-Methyl-2-butanol, ReagentPlus®, 99%