- Combined synthetic/CD strategy for the stereochemical assignment of the tricarballylic acid side chains of fumonisin B(1).
Combined synthetic/CD strategy for the stereochemical assignment of the tricarballylic acid side chains of fumonisin B(1).
The Journal of organic chemistry (2001-05-26)
M Hartl, H U Humpf
PMID11374984
ABSTRAKT
The circular dichroism (CD) exciton chirality method was employed for the stereochemical assignment of the tricarballylic acid (TCA) side chains of fumonisin B(1) 1a (FB(1)). Using 2-naphthoate for chromophoric derivatization of the reduced TCA moieties, the absolute configuration was shown to be R. For additional confirmation, an optically active dihydroxy-tert-butanoate 2 related to the TCA group of fumonisin B(1) was synthesized to serve as a model compound.
MATERIAŁY