- Potassium superoxide as an alternative reagent for Winterfeldt oxidation of beta-carbolines.
Potassium superoxide as an alternative reagent for Winterfeldt oxidation of beta-carbolines.
Organic letters (2003-01-03)
Weiqin Jiang, Xuqing Zhang, Zhihua Sui
PMID12509886
ABSTRAKT
Potassium superoxide was examined as an alternative oxidation reagent for the Winterfeldt reaction. KO(2) was found to be superior to the original Winterfeldt protocol for base-sensitive substrates. [reaction--see text]
MATERIAŁY