Merck
Molecular probe for enzymatic activity with dual output.
Bioorganic & medicinal chemistry (2007-09-18)
Eyal Danieli, Doron Shabat
ABSTRAKT
A novel molecular probe for enzymatic activity with a dual output detection-mode has been developed. The probe effectively detected the presence of the bacterial protease penicillin-G-amidase; a single cleavage by the enzyme initiated the fragmentation of a self immolative dendritic platform to release two reporter units. The signals of the free reporters were detected by two different spectroscopic techniques, fluorescence and UV-vis. This is the first reported molecular probe with two different chromogenic reporter units activated by a specific stimulus.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
6-Aminoquinoline, 98%