Merck
[Comparative research of the influence factors of DNA extraction of bloodstain on the filter paper with Chelex-100 method].
Fa yi xue za zhi (2008-01-08)
Hua-Jie Ba, Bing-quan Liu, Jun Ma, Ai-hua Zhu, Zi-qing Lin
ABSTRAKT
To improve DNA extraction from bloodstain on the filter paper and to establish a rapid, simple, and cost-effective method for DNA extraction suitable for database construction. Seven hundred and fifty two aged bloodstains on filter paper were randomly divided into four groups. The four different DNA extraction methods were compared with each other, and two DNA extraction methods used for 63 fresh bloodstains on filter paper were also compared with each other. There were no statistically significant differences observed among the four DNA extraction methods (P > 0.05) for aged bloodstains on filter paper; But the difference between the two DNA extraction methods for fresh bloodstains on filter paper was obviously (P < 0.05). Extraction of DNA samples from aged bloodstains on filter paper can be accomplished by using Chlex-100 methodology directly with no need to wash the bloodstains.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chelex® 100 sodium form, 50-100 mesh (dry)
Sigma-Aldrich
Chelex® 100 sodium form, 200-400 mesh particle size
Sigma-Aldrich
Chelex® 100 sodium form, 100-200 mesh particle size