Merck
The mismatch repair-mediated cell cycle checkpoint response to fluorodeoxyuridine.
Journal of cellular biochemistry (2008-06-11)
Angen Liu, Ken-Ichi Yoshioka, Vincenzo Salerno, Peggy Hsieh
ABSTRAKT
The loss of DNA mismatch repair (MMR) is responsible for hereditary nonpolyposis colorectal cancer and a subset of sporadic tumors. Acquired resistance or tolerance to some anti-cancer drugs occurs when MMR function is impaired. 5-Fluorouracil (FU), an anti-cancer drug used in the treatment of advanced colorectal and other cancers, and its metabolites are incorporated into RNA and DNA and inhibit thymidylate synthase resulting in depletion of dTTP and incorporation in DNA of uracil. Although the MMR deficiency has been implicated in tolerance to FU, the mechanism of cell killing remains unclear. Here, we examine the cellular response to fluorodeoxyuridine (FdU) and the role of the MMR system. After brief exposure of cells to low doses of FdU, MMR mediates DNA damage signaling during S-phase and triggers arrest in G2/M in the first cell cycle in a manner requiring MutSalpha, MutLalpha, and DNA replication. Cell cycle arrest is mediated by ATR kinase and results in phosphorylation of Chk1 and SMC1. MutSalpha binds FdU:G mispairs in vitro consistent with its being a DNA damage sensor. Prolonged treatment with FdU results in an irreversible arrest in G2 that is independent of MMR status and leads to the accumulation of DNA lesions that are targeted by the base excision repair (BER) pathway. Thus, MMR can act as a direct sensor of FdU-mediated DNA lesions eliciting cell cycle arrest via the ATR/Chk1 pathway. However, at higher levels of damage, other damage surveillance pathways such as BER also play important roles.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methoxyamine hydrochloride, 98%