Merck
[Study of DNA examination in trace sample by carrier method].
Fa yi xue za zhi (2009-02-27)
Kun-Yun Mao, Jian-Qing Zhou, Xi-Meng Gu, Hong-Yan Jiang, Yu Wang, Jia-Jia Chen
ABSTRAKT
To solve the problems of DNA testing in trace sample. Applying original carrier method to detect known trace blood DNA and to compare it with the results obtained by high effective DNA extracting method of Chelex-100. The carrier method not only could obtain the right STR genotype in the trace blood sample, but also was twice as sensitive as the Chelex-100 method. The carrier method could improve the DNA detection in trace sample. It is easy to operate and is much more valuable in the forensic case analysis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chelex® 100 sodium form, 200-400 mesh particle size
Sigma-Aldrich
Chelex® 100 sodium form, 100-200 mesh particle size
Sigma-Aldrich
Chelex® 100 sodium form, 50-100 mesh (dry)