Merck
  • PPh3-catalyzed synthesis of dicyano-2-methylenebut-3-enoates as efficient dienes in catalytic asymmetric inverse-electron-demand Diels-Alder reaction.
PPh3-catalyzed synthesis of dicyano-2-methylenebut-3-enoates as efficient dienes in catalytic asymmetric inverse-electron-demand Diels-Alder reaction.
Chemical communications (Cambridge, England) (2011-06-23)
Xianxing Jiang, Dan Fu, Xiaomei Shi, Shoulei Wang, Rui Wang
ABSTRAKT
We present here the synthesis of dicyano-2-methylenebut-3-enoates as novel Diels-Alder dienes through an efficient PPh(3)-catalyzed strategy, and an unprecedented PPh(3)-catalyzed addition/all-carbon-based asymmetric inverse-electron-demand Diels-Alder sequence reaction is disclosed for the first time.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Malononitrile, Arxada quality, ≥99.0% (calculated, GC, KF)
Sigma-Aldrich
Malononitrile, ≥99%