Merck
Suzuki-Miyaura coupling of heteroaryl boronic acids and vinyl chlorides.
Chemical communications (Cambridge, England) (2011-11-16)
Ashish Thakur, Kainan Zhang, Janis Louie
ABSTRAKT
A protocol for the Suzuki-Miyaura coupling of heteroaryl boronic acids and vinyl chlorides that minimizes protodeboronation is described. A combination of catalytic amounts of Pd(OAc)(2) and SPhos in conjunction with CsF in isopropanol effectively affords a variety of coupled products. Surprisingly, a dramatic temperature dependence in product selectivity was observed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Vinyl chloride solution, certified reference material, 2000 μg/mL in methanol
Supelco
Vinyl chloride solution, 200 μg/mL in methanol, analytical standard